Learn More
Research Products International Corp Autoclave Basket, 6 1/2 x 6 1/4 x 6 1/2 Inches, 6 per Case
SDP
Catalog No. 50212579
Encompass
Ideal for rinsing or draining glassware. Works great for moving small objects in and out of an autoclave. Can also be used for storage of test tubes or culture tubes inside incubators. All polypropylene construction will not corrode or stain metal items or glassware.
Catalog No. 50-212-579 Supplier Research Products International Corp Supplier No. 248645
Ideal for rinsing or draining glassware. Works great for moving small objects in and out of an autoclave. Can also be used for storage of test tubes or culture tubes inside incubators. All polypropylene construction will not corrode or stain metal items or glassware.