Learn More
Electron Microscopy Sciences EMS BioClave™ 16 Autoclave 16L capacity, 120 V
SDP
Catalog No. 501927906
Encompass_Preferred
Despite the space-saving exterior dimensions, the stainless steel chambers are surprisingly large, accommodating a variety of liquids, media, instruments, glassware, plasticware and other common research laboratory items. Designed specifically for non-medical research use. Includes a mechanical and electrical safety interlock that prevents the door from being opened until the pressure has reached zero PSI.
With a large digital display and fully automatic operation, all segments of the cycle (fill, sterilize, exhaust and dry) commence and run to completion by simply pressing the start button. Features a built-in water tank that does not require an external water supply. The BioClave 16 has has preset cycle options for liquids, wrapped or unwrapped instruments and an option to add drying time to end of cycle.
Volume: 16L
Catalog No. 50-192-7906 Supplier Electron Microscopy Sciences Supplier No. 8703201
Add to Cart
Add to Cart
Request a Quote
Despite the space-saving exterior dimensions, the stainless steel chambers are surprisingly large, accommodating a variety of liquids, media, instruments, glassware, plasticware and other common research laboratory items. Designed specifically for non-medical research use. Includes a mechanical and electrical safety interlock that prevents the door from being opened until the pressure has reached zero PSI.
With a large digital display and fully automatic operation, all segments of the cycle (fill, sterilize, exhaust and dry) commence and run to completion by simply pressing the start button. Features a built-in water tank that does not require an external water supply. The BioClave 16 has has preset cycle options for liquids, wrapped or unwrapped instruments and an option to add drying time to end of cycle.
Volume: 16L