Learn More
Electron Microscopy Sciences EMS BioClave™ 16 Autoclave 16L capacity, 230 V
SDP
Catalog No. 501927907
Encompass_Preferred
Despite the space-saving exterior dimensions, the stainless steel chambers are surprisingly large, accommodating a variety of liquids, media, instruments, glassware, plasticware and other common research laboratory items. Designed specifically for non-medical research use. Includes a mechanical and electrical safety interlock that prevents the door from being opened until the pressure has reached zero PSI.
With a large digital display and fully automatic operation, all segments of the cycle (fill, sterilize, exhaust and dry) commence and run to completion by simply pressing the start button. Features a built-in water tank that does not require an external water supply. The BioClave 16 has has preset cycle options for liquids, wrapped or unwrapped instruments and a option to add drying time to end of cycle.
Volume: 16L
Catalog No. 50-192-7907 Supplier Electron Microscopy Sciences Supplier No. 8703202
Add to Cart
Add to Cart
Request a Quote
Despite the space-saving exterior dimensions, the stainless steel chambers are surprisingly large, accommodating a variety of liquids, media, instruments, glassware, plasticware and other common research laboratory items. Designed specifically for non-medical research use. Includes a mechanical and electrical safety interlock that prevents the door from being opened until the pressure has reached zero PSI.
With a large digital display and fully automatic operation, all segments of the cycle (fill, sterilize, exhaust and dry) commence and run to completion by simply pressing the start button. Features a built-in water tank that does not require an external water supply. The BioClave 16 has has preset cycle options for liquids, wrapped or unwrapped instruments and a option to add drying time to end of cycle.
Volume: 16L