Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DURAN™ Autoclave Bottle with Push-On Cap
SDP

Catalog No. 14831627 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
2000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
14-831-627 1000 mL
14-831-626 500 mL
14-831-624 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-831-627 Supplier DURAN™ Supplier No. 219945452
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provides safe, convenient steam sterilization of liquid media without adjusting closures before or after autoclaving

  • Bottles are molded from DURAN™ borosilicate glass 3.3, offering outstanding thermal and chemical resistance
  • Specially designed bottle neck fits a self-venting autoclave cap for safe sterilization, with a push-on natural rubber cap for secure, easy sealing
  • Autoclavable at 121°C (slow exhaust) for reliable steam sterilization; DURAN™ sealing cap is reusable for up to five cycles
  • Reduces operator risk by eliminating manual screw cap closure steps during post-autoclave handling
  • Minimizes contamination risk by reducing manual intervention
  • Improves efficiency by eliminating manual screw cap tightening during autoclaving
  • Slim design and ergonomic grip zones enhance space utilization and handling (applies to 500 mL and 1000 mL bottles)
  • Optional labels and color-coded bottle tags enable clear lab identification (sold separately)
  • Available in matched quantities to streamline ordering and inventory management
  • 500 mL and 1000 mL bottles are sold as 4 bottles with 4 caps; 2000 mL bottles are sold as 1 bottle with 4 caps
  • Suitable substitutes for AUTOPOUR NC003, NC611, and NC608
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
Cleaning Procedure Autoclave
Color Clear
Diameter (English) Inner Neck 1.28 in.
Diameter (Metric) Inner Neck 32.5 mm
For Use With (Application) Steam sterilization, liquid media and reagents storage
Material Borosilicate Glass (3.3)
No. per Case 4/CS.
Sterility Non-sterile
Closure Color Green
Neck Finish Tapered Crimp
Closure Included Yes
Graduated Yes
Mouth Wide
Fit for Purpose Certified Quality
Height (Metric) 253 mm
Height (English) 9.96 in.
Closure Material Natural Rubber
Closure Size 45 mm
Closure Type Push-on
Autoclavable Autoclavable
Shape Round
Disposable Reusable
Product Type Media Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.