Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sterilis Solutions Autoclave Deodorizer Capsules
SDP

Catalog No. 23777006
Change view
Click to view available options
Scent:
Lemon
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Scent
23-777-006 Lemon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-777-006 Supplier Sterilis Solutions Supplier No. 38232179
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sterilis Solutions™ Autoclave Deodorizer Capsules absorb odors and replace them with a fresh, lemon scent.

  • Lemon Scented Autoclave Deodorizer Capsules are designed to absorb odors instead of masking them during the steam sterilization process
  • These gelatin capsules keep deodorizing liquid off of hands and clothes
  • Capsules ensure accurate unit dosing with each cycle presented to the Sterilis Solutions system

Specifications

Scent Lemon
Appearance Yellow
Autoclavable Autoclavable
For Use With (Application) Steam Sterilization
Quantity 50 Capsules/Jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.