Learn More
Alkali Scientific Autoclave Disposal Bags with Biohazard Print, 2mil Thickness with Temperature Indicator Strip 15.7 x 23.6", 1000/Case Clear
SDP
Catalog No. 5000663081
Encompass_Preferred
Autoclave disposal bags are designed for the safe handling and sterilization of biohazardous laboratory waste. Manufactured with a durable 2 mil thickness, these bags provide reliable strength and puncture resistance during filling, transport, and autoclaving. The biohazard symbol printed on the surface allows for easy identification and proper waste classification. Each bag includes a temperature indicator strip that visually confirms exposure to appropriate sterilization conditions. These features help ensure effective decontamination before disposal. The bags are commonly used in laboratories, healthcare facilities, and industrial settings where proper biohazard waste management is essential.
Catalog No. 50-006-63081 Supplier Alkali Scientific Supplier No. ACB16X24
Autoclave disposal bags are designed for the safe handling and sterilization of biohazardous laboratory waste. Manufactured with a durable 2 mil thickness, these bags provide reliable strength and puncture resistance during filling, transport, and autoclaving. The biohazard symbol printed on the surface allows for easy identification and proper waste classification. Each bag includes a temperature indicator strip that visually confirms exposure to appropriate sterilization conditions. These features help ensure effective decontamination before disposal. The bags are commonly used in laboratories, healthcare facilities, and industrial settings where proper biohazard waste management is essential.