Learn More
Alkali Scientific Autoclave Disposal Bags with Biohazard Print, 2mil Thickness with Temperature Indicator Strip 19.7 x 29.5", 500/Case Clear
SDP
Catalog No. 5000662737
Encompass_Preferred
Autoclave disposal bags are designed for efficient containment and sterilization of biohazardous waste in laboratory and clinical environments. Made from durable 2 mil thick material, these bags provide excellent resistance to tearing and punctures during handling and high-temperature autoclaving. The clearly printed biohazard symbol supports proper identification and safe handling procedures. Each bag is equipped with a temperature indicator strip that changes color when sterilization conditions are met, ensuring reliable verification of autoclave cycles. These bags are widely used for collecting contaminated materials and ensuring safe disposal in compliance with laboratory safety standards.
Catalog No. 50-006-62737 Supplier Alkali Scientific Supplier No. ACB20X30
Autoclave disposal bags are designed for efficient containment and sterilization of biohazardous waste in laboratory and clinical environments. Made from durable 2 mil thick material, these bags provide excellent resistance to tearing and punctures during handling and high-temperature autoclaving. The clearly printed biohazard symbol supports proper identification and safe handling procedures. Each bag is equipped with a temperature indicator strip that changes color when sterilization conditions are met, ensuring reliable verification of autoclave cycles. These bags are widely used for collecting contaminated materials and ensuring safe disposal in compliance with laboratory safety standards.