Learn More
Alkali Scientific Autoclave Disposal Bags with Biohazard Print, 2mil Thickness with Temperature Indicator Strip 31.5 x 39.4", 250/Case Clear
SDP
Catalog No. 5000662688
Encompass_Preferred
Autoclave disposal bags are designed for handling large volumes of biohazardous waste in laboratory and industrial environments. The durable 2 mil construction provides strength and puncture resistance, ensuring safe containment during collection and sterilization processes. Each bag features a clearly printed biohazard symbol to support proper identification and regulatory compliance. A temperature indicator strip is included to visually confirm that the required autoclave conditions have been reached. These bags are ideal for high-capacity waste management applications where reliable containment and effective sterilization are essential before final disposal.
Catalog No. 50-006-62688 Supplier Alkali Scientific Supplier No. AXB32X40
Autoclave disposal bags are designed for handling large volumes of biohazardous waste in laboratory and industrial environments. The durable 2 mil construction provides strength and puncture resistance, ensuring safe containment during collection and sterilization processes. Each bag features a clearly printed biohazard symbol to support proper identification and regulatory compliance. A temperature indicator strip is included to visually confirm that the required autoclave conditions have been reached. These bags are ideal for high-capacity waste management applications where reliable containment and effective sterilization are essential before final disposal.