Learn More
Alkali Scientific Autoclave Disposal Bags with Biohazard Print, 2mil Thickness with Temperature Indicator Strip 7.9 x 11.8", 1000/Case Clear
SDP
Catalog No. 5000662612
Encompass_Preferred
Autoclave disposal bags are designed for the safe collection, containment, and sterilization of biohazardous laboratory waste. These bags feature a durable 2 mil thickness that provides strength and puncture resistance during handling and autoclaving processes. The printed biohazard symbol ensures clear identification of hazardous contents, supporting compliance with laboratory safety protocols. Each bag includes a temperature indicator strip that changes color when proper sterilization conditions are reached, providing visual confirmation of successful autoclave processing. These bags are suitable for use in research laboratories, clinical settings, and industrial environments where safe disposal of contaminated materials is required.
Catalog No. 50-006-62612 Supplier Alkali Scientific Supplier No. ACB8X12
Autoclave disposal bags are designed for the safe collection, containment, and sterilization of biohazardous laboratory waste. These bags feature a durable 2 mil thickness that provides strength and puncture resistance during handling and autoclaving processes. The printed biohazard symbol ensures clear identification of hazardous contents, supporting compliance with laboratory safety protocols. Each bag includes a temperature indicator strip that changes color when proper sterilization conditions are reached, providing visual confirmation of successful autoclave processing. These bags are suitable for use in research laboratories, clinical settings, and industrial environments where safe disposal of contaminated materials is required.