Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Clavies™ Heat-Resistant Biohazard Autoclave Gloves
SCI_ED

Catalog No. S47334 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Length (English) Cuff:
5 in.
11 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Cuff
S47334 5 in.
S473341 11 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S47334 Supplier Bel-Art™ Supplier No. H132010000/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Soft, pliant, cotton terrycloth gloves are ideal for handling hot objects out of the autoclave or oven and protect from temperatures up to 450°F (232°C).

  • Ideal for removing hot objects from an autoclave or oven
  • Heat-resistant up to 450°F (232°C)
  • Replaces hazardous asbestos gloves
  • Extended gauntlet cuff available for additional forearm protection
  • Gloves are not waterproof and should not be used to handle dry ice
  • Machine wash, air dry, or use the Lab-Aire™ II Glove Holder (F18931-0000) to dry on any Lab-Aire™ dryer

Specifications

Certifications/Compliance CE
Accelerator Free Yes
Applications Laboratory, Industrial
Coating Material Uncoated
Color Orange
Latex Free Yes
Material Terrycloth
Product Type Autoclave Gloves
Size Universal
Style Full-hand Mitt
Temperature (English) Max. 450°F
Temperature (Metric) Max 232°C
For Use With (Application) Handling Hot Objects
Glove Cuff Style Gauntlet
Series H13201
Length (English) 13 in.
Length (English) Cuff 5 in.
Length (Metric) 33 cm
Length (Metric) Cuff 13 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.