Learn More
Bel-Art™ Clavies™ Heat-Resistant Biohazard Autoclave Gloves
Shop All SP Bel-Art & SP Wilmad Products

Description
- Ideal for removing hot objects from an autoclave or oven
- Heat-resistant up to 450°F (232°C)
- Replaces hazardous asbestos gloves
- Extended gauntlet cuff available for additional forearm protection
- Gloves are not waterproof and should not be used to handle dry ice
- Machine wash, air dry, or use the Lab-Aire™ II Glove Holder (F18931-0000) to dry on any Lab-Aire™ dryer
Specifications
Specifications
| Certifications/Compliance | CE |
| Accelerator Free | Yes |
| Applications | Laboratory, Industrial |
| Coating Material | Uncoated |
| Color | Orange |
| Latex Free | Yes |
| Material | Terrycloth |
| Product Type | Autoclave Gloves |
| Size | Universal |
| Style | Full-hand Mitt |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.