Learn More
Sigma Aldrich Fine Chemicals Biosciences Autoclave pans capacity 3.1EA
Catalog No. 502900620
Encompass
Autoclavable polypropylene with strong rims. Sizes for small instruments and labware. Nestable when empty. Without handles.
Catalog No. 50-290-0620 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z3603921EA
Autoclavable polypropylene with strong rims. Sizes for small instruments and labware. Nestable when empty. Without handles.