Learn More
Sigma Aldrich Fine Chemicals Biosciences Autoclave pans capacity 51EA
Catalog No. 502901309
Encompass
Autoclavable polypropylene with strong rims. Sizes for small instruments and labware. Nestable when empty. Without handles.
Catalog No. 50-290-1309 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z3604061EA
Autoclavable polypropylene with strong rims. Sizes for small instruments and labware. Nestable when empty. Without handles.