Learn More
Electron Microscopy Sciences Autoclave Tape Printed Tape "Sterilized" Darkens and Appears, 3/4" width 60 YD
SDP
Catalog No. 501930834
Encompass_Preferred
Versatile imprinted tape indicates completion of normal steam sterilization processing. Tape accepts handwritten notations, may be used to seal and identify laboratory materials before and after processing. "Sterilized" darkens in color. Tape is pre-printed with "Batch", "Con.#" and "Date".
Width: 3/4"
Catalog No. 50-193-0834 Supplier Electron Microscopy Sciences Supplier No. 7701807
Versatile imprinted tape indicates completion of normal steam sterilization processing. Tape accepts handwritten notations, may be used to seal and identify laboratory materials before and after processing. "Sterilized" darkens in color. Tape is pre-printed with "Batch", "Con.#" and "Date".
Width: 3/4"