Learn More
GA International Autoclave-Resistant Labels for Inkjet Printers 3.5" x 2" / 88.9mm x 50.8mm Rectangle White From -40°C to +150°C (-40°F to +302°F)
Catalog No. NC1858335
Encompass_Preferred
Permanent inkjet printable labels, ideal for glassware, metal and plastic surfaces, trays, and other items intended for use in steam autoclaves, ovens, and gamma irradiators (tested up to 50 KGy). Our autoclave inkjet printable labels are durable, weather & water-proof, alcohol-resistant, and have been tested to endure 10 autoclave cycles (up to 121°C/250°F, 16.5 psi for 20 minutes). Compatible with all major brands of inkjet printers and are easy to format for use with our free online Microsoft Word templates.
Catalog No. NC1858335 Supplier GA International Supplier No. INKA61WH
Permanent inkjet printable labels, ideal for glassware, metal and plastic surfaces, trays, and other items intended for use in steam autoclaves, ovens, and gamma irradiators (tested up to 50 KGy). Our autoclave inkjet printable labels are durable, weather & water-proof, alcohol-resistant, and have been tested to endure 10 autoclave cycles (up to 121°C/250°F, 16.5 psi for 20 minutes). Compatible with all major brands of inkjet printers and are easy to format for use with our free online Microsoft Word templates.