Learn More
SPS Medical Tape Dispenser 1 Inch - 1 Count
Catalog No. nc0018358
Encompass
Indicator tapes provide immediate identification of processed items while also securing sterilization packs. Color change indicator verifies exposure to sterilization process. Tapes are individually packaged in 60 yard rolls and available in multiple widths. Blue steam tape has been specially designed with aggressive adhesive for disposable CSR wraps. Tape dispenser can hold up to 2 rolls of 1 tape and has a weighted base.
Catalog No. NC0018358 Supplier SPS Medical Supplier No. TD001
Indicator tapes provide immediate identification of processed items while also securing sterilization packs. Color change indicator verifies exposure to sterilization process. Tapes are individually packaged in 60 yard rolls and available in multiple widths. Blue steam tape has been specially designed with aggressive adhesive for disposable CSR wraps. Tape dispenser can hold up to 2 rolls of 1 tape and has a weighted base.