Learn More
Thermco Products Inc AUTOCLAVE THERMOMETER NIST
Catalog No. NC0700040
Encompass_Preferred
Maximum registering thermometer used for the verification of the temperature in autoclaves and sterilizers. The thermometer will indicate the highest temperature in such instruments. After each use the thermometer must be shaken down to reset the thermometer before being used again. The ACC80135SC is safety-coated with PTFE, so no glass or mercury will be emited if broken. Can be certified treceable to N.I.S.T. individually serialized. The ACC80135SCA is housed in an eight inch nickel plated brass metal armer with a slotted window for extra protection.
Catalog No. NC0700040 Supplier Thermco Products Inc Supplier No. ACC80135SCA
Maximum registering thermometer used for the verification of the temperature in autoclaves and sterilizers. The thermometer will indicate the highest temperature in such instruments. After each use the thermometer must be shaken down to reset the thermometer before being used again. The ACC80135SC is safety-coated with PTFE, so no glass or mercury will be emited if broken. Can be certified treceable to N.I.S.T. individually serialized. The ACC80135SCA is housed in an eight inch nickel plated brass metal armer with a slotted window for extra protection.