Learn More
Thermco Products Inc Armored Thermometer Certified at 120°C for Dairies
Catalog No. NC1886013
Encompass_Preferred
Autoclave Maximum Registering Thermometers are for validating autoclaves by monitoring the highest temperature attained during a sterilization cycle. These thermometers operate on the same principle as thermometers designed to measure fevers: the highest temperature attained is recorded, and doesn’t recede until the thermometer is shaken down.
Catalog No. NC1886013 Supplier Thermco Products Inc Supplier No. ACC80135DSCFCA
Autoclave Maximum Registering Thermometers are for validating autoclaves by monitoring the highest temperature attained during a sterilization cycle. These thermometers operate on the same principle as thermometers designed to measure fevers: the highest temperature attained is recorded, and doesn’t recede until the thermometer is shaken down.