Learn More
Research Products International Corp Autoclavable Plastic Tubing Dispenser, 12 Wide
SDP
Catalog No. 50212683
Encompass
Create custom size bags for hybridization procedures or for autoclaving individual items. Ultra-clear 2 ml polypropylene tubing allows full visibility of bag contents. Tubing can be sealed quickly using an electric bag sealer. Optional dispenser provides a convenient method for keeping tubing on a benchtop or can be wall mounted. Divider kit organizes various size rolls for convenient dispensing.
Catalog No. 50-212-683 Supplier Research Products International Corp Supplier No. 274561
Create custom size bags for hybridization procedures or for autoclaving individual items. Ultra-clear 2 ml polypropylene tubing allows full visibility of bag contents. Tubing can be sealed quickly using an electric bag sealer. Optional dispenser provides a convenient method for keeping tubing on a benchtop or can be wall mounted. Divider kit organizes various size rolls for convenient dispensing.