Learn More
Cole-Parmer Thermo Scientific Nalgene 6902-5000 polypropylene autoclaving tray, 5 L
Catalog No. NC9257661 Shop All Cole-Parmer Products
Encompass_Preferred
Polypropylene (PP) trays feature a tapered design for nesting and rolled edges for strength. Autoclavable. NALGENE. Pack of 3.
Catalog No. NC9257661 Supplier Cole-Parmer Supplier No. 0601023
Polypropylene (PP) trays feature a tapered design for nesting and rolled edges for strength. Autoclavable. NALGENE. Pack of 3.