Learn More
Alzet AutoClip Applier for 9 mm Wound Clips
SDP
Catalog No. NC0434080
Encompass
The AutoClip Applier is designed for use with 9 mm AutoClips as part of the AutoClip wound closure system, which provides a fast and effective alternative to sutures for closing wounds in research animals. The system is ideal for quickly closing skin incisions made for ALZET pump implantations, enabling researchers to streamline surgeries and save valuable time. The automatic, spring-loaded AutoClip Applier places 9 mm AutoClips (sold separately) such that they provide maximum holding force while minimizing skin trauma. After the incision is healed, the wound clips are easily removed using the AutoClip Remover (sold separately). The AutoClip Applier is supplied non-sterile but may be autoclaved.[AutoClip 9 mm wound clips and the AutoClip Remover are sold separately and can be purchased individually or as part of several kits.]
Catalog No. NC0434080 Supplier Alzet Supplier No. 0009956
The AutoClip Applier is designed for use with 9 mm AutoClips as part of the AutoClip wound closure system, which provides a fast and effective alternative to sutures for closing wounds in research animals. The system is ideal for quickly closing skin incisions made for ALZET pump implantations, enabling researchers to streamline surgeries and save valuable time. The automatic, spring-loaded AutoClip Applier places 9 mm AutoClips (sold separately) such that they provide maximum holding force while minimizing skin trauma. After the incision is healed, the wound clips are easily removed using the AutoClip Remover (sold separately). The AutoClip Applier is supplied non-sterile but may be autoclaved.[AutoClip 9 mm wound clips and the AutoClip Remover are sold separately and can be purchased individually or as part of several kits.]