Learn More
Stoelting™ Autoclip Applier and Accessories
SDP
Catalog No. 10001032 Shop All Stoelting Co Products
Change view
Click to view available options
Includes:
Applier
Autoclips
Remover
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
10-001-032 Autoclips
10-001-030 Applier
10-001-031 Remover
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-001-032 Supplier Stoelting™ Supplier No. 59047
Add to Cart
Add to Cart
Autoclip™ Applier works like a staple gun to rapidly apply clip closures.
  • Applier holds 20, 9mm clips, which are held firm and ready for use by rustless beryllium-copper springs
  • Provides maximum holding force with minimal trauma
  • No need for assistance to operate the applier or remover
  • When the wound is healed, Clips are easily removed with the Autoclip or EZ Clip Remover.
Specifications
Model Autoclip
Quantity 1000/Cs.
Includes Autoclips