Learn More
Alzet AutoClip Remover for 9 mm Wound Clips
SDP
Catalog No. NC0477695
Encompass
The AutoClip Remover is designed for use with 9 mm AutoClips as part of the AutoClip wound closure system, which provides a fast and effective alternative to sutures for closing wounds in research animals. The system is ideal for quickly closing skin incisions made for ALZET pump implantations, enabling researchers to streamline surgeries and save valuable time. After wound healing, the AutoClip Remover is used to remove the 9 mm AutoClips (sold separately) which were initially placed using the AutoClip Applier (sold separately). The AutoClip Remover is supplied non-sterile but may be autoclaved.[AutoClip 9 mm wound clips and the AutoClip Applier are sold separately and can be purchased individually or as part of several kits.]
Catalog No. NC0477695 Supplier Alzet Supplier No. 0009957
The AutoClip Remover is designed for use with 9 mm AutoClips as part of the AutoClip wound closure system, which provides a fast and effective alternative to sutures for closing wounds in research animals. The system is ideal for quickly closing skin incisions made for ALZET pump implantations, enabling researchers to streamline surgeries and save valuable time. After wound healing, the AutoClip Remover is used to remove the 9 mm AutoClips (sold separately) which were initially placed using the AutoClip Applier (sold separately). The AutoClip Remover is supplied non-sterile but may be autoclaved.[AutoClip 9 mm wound clips and the AutoClip Applier are sold separately and can be purchased individually or as part of several kits.]