Learn More
Roboz Surgical AUTOCLIP REMOVING FCPS 4IN
SDP
Catalog No. NC9466048
Encompass
Reflex Wound Clip Remover; For Use With Reflex 9mm And 7mm Clips, And Other Brands; 4 in. Length
Catalog No. NC9466048 Supplier Roboz Surgical Supplier No. RS9263
Reflex Wound Clip Remover; For Use With Reflex 9mm And 7mm Clips, And Other Brands; 4 in. Length