Learn More
Fine Science Tools AutoClip System, Secure Alternative Method of Wound Closure
SDP
Catalog No. NC0958957
Encompass
The AutoClip System is a safe, secure alternative method of wound closure. Faster and easier than suture, this system is perfect when staples may be too thin to provide adequate closure. Components sold separately or as a kit. Kit includes: 1 Applier, 9mm (No. 12020-09); 100 Wound clips, 9mm (No. 12022-09); 1 Remover (No. 12023-00)
Catalog No. NC0958957 Supplier Fine Science Tools Supplier No. 1202000
The AutoClip System is a safe, secure alternative method of wound closure. Faster and easier than suture, this system is perfect when staples may be too thin to provide adequate closure. Components sold separately or as a kit. Kit includes: 1 Applier, 9mm (No. 12020-09); 100 Wound clips, 9mm (No. 12022-09); 1 Remover (No. 12023-00)