Learn More
Braintree Scientific 9mm Autoclip, Kit, Applier/Remover/1,000 Staples
SDP
Catalog No. NC9946451
Encompass
Autoclip Wound Closing System - The Autoclip Applier works like a staple gun to rapidly apply Autoclip closures. The applier will hold 20 Autoclips, which are held firm and ready for use by rustless beryllium-copper springs. Autoclips provide maximum holding force with minimal trauma. There is no need for assistance to operate the applier or remover. When the wound is healed, clips are easily removed with the Autoclip Remover. Stainless steel Autoclips are non-pyrogenic and supplied non-sterile, but may be autoclaved. Ideal for use with laboratory mice and rats. **Sold for research purposes only and not for human use**
Catalog No. NC9946451 Supplier Braintree Scientific Supplier No. ACSKIT
 Restricted Item
Add to Cart
Add to Cart
Autoclip Wound Closing System - The Autoclip Applier works like a staple gun to rapidly apply Autoclip closures. The applier will hold 20 Autoclips, which are held firm and ready for use by rustless beryllium-copper springs. Autoclips provide maximum holding force with minimal trauma. There is no need for assistance to operate the applier or remover. When the wound is healed, clips are easily removed with the Autoclip Remover. Stainless steel Autoclips are non-pyrogenic and supplied non-sterile, but may be autoclaved. Ideal for use with laboratory mice and rats. **Sold for research purposes only and not for human use**