Learn More
Stoelting™ Autoclip Wound Closure Kit
SDP
Catalog No. 10001029 Shop All Stoelting Co Products
Change view
Click to view available options
For Use With (Application):
Neuroscience
Catalog No. For Use With (Application)
10-001-029 Neuroscience
1 options
Catalog No. 10-001-029 Supplier Stoelting™ Supplier No. 59040
Add to Cart
Add to Cart
Stoelting™ Autoclip Wound Closure Kit works like a staple gun to rapidly apply clip closures.
  • Applier will hold 20, 9 mm clips, which are held firm and ready for use by rustless beryllium-copper springs
  • Provides maximum holding force with minimal trauma.
  • No need for assistance to operate the applier or remover.
  • When the wound is healed, clips are easily removed with the Autoclip or EZ Clip Remover
Specifications
For Use With (Application) Neuroscience