Learn More
Electron Microscopy Sciences Autoclave Indicator Tape | 19mm x 54.9m (1 Roll)
SDP
Catalog No. 501927386
Encompass_Preferred
This autoclave Indicator tape has printed diagonal stripes that darken indicating a steam autoclave process is complete. Stripes darken after 5 minutes of exposure at 273°F (134°C) at steam pressure of 1.02 kg/cm2 or marker.
Catalog No. 50-192-7386 Supplier Electron Microscopy Sciences Supplier No. 7701620
This autoclave Indicator tape has printed diagonal stripes that darken indicating a steam autoclave process is complete. Stripes darken after 5 minutes of exposure at 273°F (134°C) at steam pressure of 1.02 kg/cm2 or marker.