Learn More
Electron Microscopy Sciences Autoclave Indicator Tape, 1.00" wide 2160 IN (1 Roll)
SDP
Catalog No. 501927387
Encompass_Preferred
Printed diagonal stripes darken indicating a steam autoclave process is complete. Stripes darken after 5 minutes of exposure at 273 degree F (134 degree C) at steam pressure of 1.02 kg/cm2 or marker.Width: 1.00"
Catalog No. 50-192-7387 Supplier Electron Microscopy Sciences Supplier No. 7701621
Printed diagonal stripes darken indicating a steam autoclave process is complete. Stripes darken after 5 minutes of exposure at 273 degree F (134 degree C) at steam pressure of 1.02 kg/cm2 or marker.Width: 1.00"