Learn More
Foxx Life Sciences Autofil 2 Bottle Top Vacuum Filtration unit, Funnel Only, 1000 mL, 0.22 µm PES Membrane, Sterile, 12/CS
GREENER_CHOICE
Catalog No. 5000574393
Encompass_Preferred
The Autofil 2 Bottle Top Vacuum Filter Funnel Only features 0.1um, 0.2um, or 0.45um PES membranes. Perfect for filtering FBS and other viscous solutions. The membrane is low protein binding and surfactant-free.
Features
Available in 250ml, 500ml and 1000ml. Funnel is compatible with GL 45 thread bottles
Bottle top Filter units include adapter for connecting to vacuum hose
Design includes the NEW EZLabpure™ ergonomic bottle shape with molded finger grip
GL 45 caps provides a convenient gripping surface
All units are sterile and manufactured from USP Class VI polystyrene in an ISO 13485 clean room
Catalog No. 50-005-74393 Supplier Foxx Life Sciences Supplier No. 11532FLS
The Autofil 2 Bottle Top Vacuum Filter Funnel Only features 0.1um, 0.2um, or 0.45um PES membranes. Perfect for filtering FBS and other viscous solutions. The membrane is low protein binding and surfactant-free.
Features
Available in 250ml, 500ml and 1000ml. Funnel is compatible with GL 45 thread bottles
Bottle top Filter units include adapter for connecting to vacuum hose
Design includes the NEW EZLabpure™ ergonomic bottle shape with molded finger grip
GL 45 caps provides a convenient gripping surface
All units are sterile and manufactured from USP Class VI polystyrene in an ISO 13485 clean room