Learn More
Foxx Life Sciences Autofil Sterile Disposable Centrifuge Tube Vacuum Filter Stand, For 15mL and 50mL Tubes
Catalog No. 501178701
Encompass_Preferred
146-9004-RLS: Centrifuge Tube Stand, 15ml and 50 ml. Foxx Life Sciences' Centrifuge Tube Stands are intended for use with centrifuge tubes. They are made from Class VI, FDA Grade, BPA-Free Polystyrene, and are conveniently sold in packs of 5. Autofil tube stand can hold one 15ml or 50ml tube. Can also be used with the Autofil Centrifuge Filtration system.
Catalog No. 50-117-8701 Supplier Foxx Life Sciences Supplier No. 1469004RLS
146-9004-RLS: Centrifuge Tube Stand, 15ml and 50 ml. Foxx Life Sciences' Centrifuge Tube Stands are intended for use with centrifuge tubes. They are made from Class VI, FDA Grade, BPA-Free Polystyrene, and are conveniently sold in packs of 5. Autofil tube stand can hold one 15ml or 50ml tube. Can also be used with the Autofil Centrifuge Filtration system.