Learn More
Foxx Life Sciences Bottle Top Vacuum Filtration Cradle Ring
Catalog No. NC2865543
Encompass_Preferred
This hands-free filtration system is designed to work seamlessly with the Quick-Connect™ Pedestal. Constructed from durable stainless steel, this patented docking system ensures long-lasting performance and convenience.
Key Benefits:
Hands-free operation for increased safety and productivity
Integrated on/off switch for precise vacuum flow control
Permanent hose connection to minimize setup time and improve workflow
Works in conjunction with FOXX APEXTM Bottle Top Filter Assembly and the EZLabpure Bottle Top Filter Assembly.
Catalog No. NC2865543 Supplier Foxx Life Sciences Supplier No. 1122RLS
This hands-free filtration system is designed to work seamlessly with the Quick-Connect™ Pedestal. Constructed from durable stainless steel, this patented docking system ensures long-lasting performance and convenience.
Key Benefits:
Hands-free operation for increased safety and productivity
Integrated on/off switch for precise vacuum flow control
Permanent hose connection to minimize setup time and improve workflow
Works in conjunction with FOXX APEXTM Bottle Top Filter Assembly and the EZLabpure Bottle Top Filter Assembly.