Learn More
GUARDIAN EQUIPMENT Eyewash, Deck Mounted AutoFlow™ 90° Swivel, Right Hand Mounting
Catalog No. NC1146858
Encompass_Preferred
AutoFlow™ eyewash for mounting on counter next to sink. Swinging the spray head assembly 90 degrees horizontally out over the sink activates the water flow. The unit remains in operation until the spray head assembly is swung back into the storage position, closing the valve. Unit is also available with left hand mounting configuration.
Catalog No. NC1146858 Supplier GUARDIAN EQUIPMENT Supplier No. G1805
AutoFlow™ eyewash for mounting on counter next to sink. Swinging the spray head assembly 90 degrees horizontally out over the sink activates the water flow. The unit remains in operation until the spray head assembly is swung back into the storage position, closing the valve. Unit is also available with left hand mounting configuration.