Learn More
MilliporeSigma AUTOFLUOR ELIMINATOR REAG 50ML
Catalog No. NC9310541
Encompass
Autofluorescence Eliminator Reagent, Ready-to-use for application in fluorescence microscopy procedures, Liquid in 70% ethanol containing no preservatives, Store at room Temperature in the dark, 50mL
Catalog No. NC9310541 Supplier MilliporeSigma Supplier No. 2160
Autofluorescence Eliminator Reagent, Ready-to-use for application in fluorescence microscopy procedures, Liquid in 70% ethanol containing no preservatives, Store at room Temperature in the dark, 50mL