Learn More
Mitegen AutoGrid Cassette Storage Puck Starter Kit with Loading Station Insert, Tongs, and Storage Cane
SDP
Catalog No. 5000495030
Encompass_Preferred
The MiTeGen AutoGrid Cassette Storage Puck Starter Kit includes all the basic components needed to begin working with AutoGrid cassettes. The kit contains a MiTeGen Autogrid Cassette Puck (M-CEM-AGC-P1), a Loading Station Insert (M-CEM-AGC-PLSI1), a pair of Cryo-EM Puck Grasping Tongs (M-CEMG2-PGT1), a Shelved Puck Storage Cane for HC34, TW34, ICB34 or VHC35 (M-CEMSTC-2), and a Puck Locking Screw Driver (M-CEMG2-LLSD).
Catalog No. 50-004-95030 Supplier Mitegen Supplier No. MCEMAGCSK1
The MiTeGen AutoGrid Cassette Storage Puck Starter Kit includes all the basic components needed to begin working with AutoGrid cassettes. The kit contains a MiTeGen Autogrid Cassette Puck (M-CEM-AGC-P1), a Loading Station Insert (M-CEM-AGC-PLSI1), a pair of Cryo-EM Puck Grasping Tongs (M-CEMG2-PGT1), a Shelved Puck Storage Cane for HC34, TW34, ICB34 or VHC35 (M-CEMSTC-2), and a Puck Locking Screw Driver (M-CEMG2-LLSD).