Learn More
Electron Microscopy Sciences AUTOGRID CMPTIBLE CRYO GRID BX
SDP
Catalog No. NC2098973
Encompass
Autogrid Compatible Cryo Grid Box. Features 4 large diamond-shaped cavities that accommodate larger diameter AutoGrids.
This item is not returnable. View return policy
Catalog No. NC2098973 Supplier Electron Microscopy Sciences Supplier No. 71166AG
Autogrid Compatible Cryo Grid Box. Features 4 large diamond-shaped cavities that accommodate larger diameter AutoGrids.