Learn More
Mitegen Autogrid Rings & C-clips By Nanosoft For Cryo-EM & Autoloader Systems, 100 Rings & 100 C-clips Pack
SDP
Catalog No. NC2752966
Encompass
Autogrid Rings and C-clips by Nanosoft provide a premium, functionally-equivalent solution for clipping grids in Thermo Fisher Scientific (TFS) cryo-EMs and autoloader systems. Manufactured using cutting-edge technology, ensuring consistent and reliable performance. They offer exceptional value as a high-quality, one-time use consumable. Extensive testing across numerous facilities confirms reliable operation without defects in autoloader or microscope functions. These C-clips are high-quality and fully compatible with both TFS autogrid rings and Nanosoft autogrid rings. The rings are also compatible with TFS C-clips. Both components are designed for seamless use with Nanosoft and TFS clipping stations and tools, optimizing your cryo-EM sample preparation workflow. This convenient 100-pack includes 100 Autogrid Rings and 100 C-clips.
Catalog No. NC2752966 Supplier Mitegen Supplier No. MCEMNS11011001
Autogrid Rings and C-clips by Nanosoft provide a premium, functionally-equivalent solution for clipping grids in Thermo Fisher Scientific (TFS) cryo-EMs and autoloader systems. Manufactured using cutting-edge technology, ensuring consistent and reliable performance. They offer exceptional value as a high-quality, one-time use consumable. Extensive testing across numerous facilities confirms reliable operation without defects in autoloader or microscope functions. These C-clips are high-quality and fully compatible with both TFS autogrid rings and Nanosoft autogrid rings. The rings are also compatible with TFS C-clips. Both components are designed for seamless use with Nanosoft and TFS clipping stations and tools, optimizing your cryo-EM sample preparation workflow. This convenient 100-pack includes 100 Autogrid Rings and 100 C-clips.