Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ AutoLys M Tube Pliers

Catalog No. A38261
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A38261 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A38261 Supplier Applied Biosystems™ Supplier No. A38261
  • $86.25
    $80.65 / Each of 1

    Save $5.60

    6% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Centrifugation of the tubes in an AutoLys M Tube Rack is required when processing FFPE samples using the MagMAX FFPE DNA/RNA Ultra Kit.

Centrifugation of the tubes in an AutoLys M Tube Rack is required when processing FFPE samples using the MagMAX FFPE DNA/RNA Ultra Kit. The outer-tube will contain the lysate that is processed further to recover the nucleic acids.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) AutoLys TubeLifter System
Quantity 1 each
Shipping Condition Room Temperature
Can I use a heat block for the AutoLys M tube incubation?

No. The AutoLys M tubes don't fit in a standard 1.5 mL or 2.0 mL heat block. We recommend incubating the tubes in the tube racks in an incubator or oven.

What kind of centrifuge do I need to spin down the AutoLys M Tubes (Cat. No. A38738)?

Any centrifuge with plate adapters will work. The AutoLys M Tube Racks fit nicely into those adapters.

Do I need to buy the AutoLys M TubeLifter (Cat. No. A37956) or AutoLys M Tube Pliers (Cat. No. A38261) to use the AutoLys M Tubes (Cat. No. A38738)?

No, the AutoLys M Tubes can be lifted manually, but the tools significantly reduce hand strain and increase ease of use as well as throughput.

Do the AutoLys M Tubes come with the MagMAX FFPE DNA/RNA Ultra Kit?

No, they have to be purchased separately together with some other products that are necessary for using the AutoLys M Tube system efficiently:
- AutoLys M Tubes and Caps (Cat. No. A38738)
- Autolys M Tube Locking Lid (Cat. No. A37954)
- AutoLys M TubeLifter (Cat. No. A37956)
- AutoLys M Tube Pliers (Cat. No. A38261)
- AutoLys M Tube Racks (Cat. No. A37955)

How does using the the AutoLys M Tube system help in increase the DNA/RNA yield from FFPE samples?

The FFPE-associated wax and debris are held in the upper chamber of the AutoLys M tube while the lysate passes through. Nucleic acids can then be purified from the clarified lysate. The AutoLys M tubes thus reduce manipulation of the sample eliminating the need to pellet the tissue and wash, so tissue loss is minimized

Can I use the AutoLys M Tube system with any of your FFPE sample prep kits or has it only been optimized for the MagMAX FFPE DNA/RNA Ultra Kit?

The AutoLys M Tube system has only been tested with the MagMAX FFPE DNA/RNA Ultra Kit, but it may also work with our RecoverAll Total Nucleic Acid Isolation Kit for FFPE .

I am planning on using your MagMAX FFPE DNA/RNA Ultra Kit. I don't want to perform a deparaffinization step using organic solvents, is there an alternative reagent or method I can use?

Yes, we offer the AutoLys M Tube system, which creates cleared lysates from FFPE tissue samples without the need for deparaffinization or wash steps, while increasing DNA and RNA yields.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.