Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ AutoLys M Tube Racks

Catalog No. A37955
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 racks
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A37955 5 racks
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A37955 Supplier Applied Biosystems™ Supplier No. A37955
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These AutoLys M Tube Racks hold 24 AutoLys M Tubes per rack.

• These AutoLys M Tube Racks hold 24 AutoLys M Tubes per rack
• Racks are locked and unlocked using the AutoLys M TubeLifter.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 5 racks (each rack holds up to 24 Tubes)
For Use With (Equipment) AutoLys TubeLifter System
Quantity 5 racks
Shipping Condition Room Temperature
Can I use a heat block for the AutoLys M tube incubation?

No. The AutoLys M tubes don't fit in a standard 1.5 mL or 2.0 mL heat block. We recommend incubating the tubes in the tube racks in an incubator or oven.

What kind of centrifuge do I need to spin down the AutoLys M Tubes (Cat. No. A38738)?

Any centrifuge with plate adapters will work. The AutoLys M Tube Racks fit nicely into those adapters.

Do I need to buy the AutoLys M TubeLifter (Cat. No. A37956) or AutoLys M Tube Pliers (Cat. No. A38261) to use the AutoLys M Tubes (Cat. No. A38738)?

No, the AutoLys M Tubes can be lifted manually, but the tools significantly reduce hand strain and increase ease of use as well as throughput.

Do the AutoLys M Tubes come with the MagMAX FFPE DNA/RNA Ultra Kit?

No, they have to be purchased separately together with some other products that are necessary for using the AutoLys M Tube system efficiently:
- AutoLys M Tubes and Caps (Cat. No. A38738)
- Autolys M Tube Locking Lid (Cat. No. A37954)
- AutoLys M TubeLifter (Cat. No. A37956)
- AutoLys M Tube Pliers (Cat. No. A38261)
- AutoLys M Tube Racks (Cat. No. A37955)

How does using the the AutoLys M Tube system help in increase the DNA/RNA yield from FFPE samples?

The FFPE-associated wax and debris are held in the upper chamber of the AutoLys M tube while the lysate passes through. Nucleic acids can then be purified from the clarified lysate. The AutoLys M tubes thus reduce manipulation of the sample eliminating the need to pellet the tissue and wash, so tissue loss is minimized

Can I use the AutoLys M Tube system with any of your FFPE sample prep kits or has it only been optimized for the MagMAX FFPE DNA/RNA Ultra Kit?

The AutoLys M Tube system has only been tested with the MagMAX FFPE DNA/RNA Ultra Kit, but it may also work with our RecoverAll Total Nucleic Acid Isolation Kit for FFPE .

I am planning on using your MagMAX FFPE DNA/RNA Ultra Kit. I don't want to perform a deparaffinization step using organic solvents, is there an alternative reagent or method I can use?

Yes, we offer the AutoLys M Tube system, which creates cleared lysates from FFPE tissue samples without the need for deparaffinization or wash steps, while increasing DNA and RNA yields.


WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.