Learn More
Yamato Scientific America, Inc. Yamato Fully-Automatic Benchtop Glassware Laboratory Washer, 115V/220V.
Catalog No. 501898203 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Compact and powerful automatic benchtop washer with spin table that helps reduce laboratory glassware cleaning workload. Automatic compact glassware washer with timer and washing program according to glassware shape and contamination. Final rinse (option) with purified water available. Water purifier connection is possible for pure water rinse process. Wash process and time can be set according to glassware shape and contamination level. Cleaning water temperature impacts final cleaning results. With built-in water heater, no boiler piping and water heating system are required. Powerful upward and downward two-way pressurized water jet method. Optional jet rack is available for hard to clean targets, such as glassware with narrow neck or body. Washing water temp. 45 ~ 80°C.
Catalog No. 50-189-8203 Supplier Yamato Scientific America, Inc. Supplier No. AW62
Add to Cart
Add to Cart
Request a Quote
Compact and powerful automatic benchtop washer with spin table that helps reduce laboratory glassware cleaning workload. Automatic compact glassware washer with timer and washing program according to glassware shape and contamination. Final rinse (option) with purified water available. Water purifier connection is possible for pure water rinse process. Wash process and time can be set according to glassware shape and contamination level. Cleaning water temperature impacts final cleaning results. With built-in water heater, no boiler piping and water heating system are required. Powerful upward and downward two-way pressurized water jet method. Optional jet rack is available for hard to clean targets, such as glassware with narrow neck or body. Washing water temp. 45 ~ 80°C.