Learn More
Beckman Coulter™ Automate 2500 Systems Recap
Catalog No. 01361059 Shop All Beckman Coulter Products
Change view
Click to view available options
Speed:
1,200 Tubes/hr.
Catalog No. Speed
01-361-059 1,200 Tubes/hr.
1 options
Catalog No. 01-361-059 Supplier Beckman Coulter™ Supplier No. A99172
Add to Cart
Add to Cart
The AutoMate family of sample processing systems allows you to streamline pre- and post-analytical processes to help you gain optimal performance and use of resources, eliminating steps between sample receipt and analysis.
When you need to improve efficiency, turn to the AutoMate 2500 family of sample processing systems. The AutoMate 2500 family features Intelligent Sample Banking (ISB) software to deliver an automation system that fits your laboratory's unique needs. The AutoMate family of sample processing systems allows you to streamline pre- and post-analytical processes to help you gain optimal performance and use of resources, eliminating steps between sample receipt and analysis. ISB enhances your automated solution by enabling you to build and manage a sample bank with maximum efficiency and security and increased control over sample traceability.
Advanced capabilities
  • Minimize manual handling with automated sample loading and sorting
  • Simplify measuring with through-the-label sample volume detection
  • Enable fast processing with sorting speeds of 800 to 1,200 tubes per hour
  • Reduce manual sample preparation errors and ensure faster, more accurate secondary tube preparation with intelligent aliquoting and tube labeling
  • Experience ease of use with intuitive software
  • Transfer sample to archiving safely and conveniently with recapper technology
Single point of entry
  • Manage sample tubes from entry into the laboratory until archiving
  • Re-sort samples for pending test routing or for storage when samples have volume remaining
  • Sort STATs to separate locations
  • Provide sorting flexibility with up to 24 dynamically configurable rack locations
  • Unload and load instrument-specific racks
Tube inspection unit cap color recognition
  • Validate sample type against test ordered to prevent errors with cap color analysis
  • Detect the newest generation of coded caps
Volume determination
  • Gain better sample control with automated measurement of each sample's volume prior to dispensing into aliquot tube or for storage
  • Ascertain whether all aliquots can be created through automated measurement of plasma or serum volume
  • Allow infrared volume detection of up to three layers of labels on the tube with read-through labels
TRUSTED_SUSTAINABILITY
Specifications
Operation Automated
Type Capper/Decapper
For Use With (Equipment) Beckman Coulter Analyzers
Speed 1,200 Tubes/hr.