Learn More
Molecular Devices Molecular Devices Imager, Brightfield and Fluorescence, 96/384-Well, High Resolution
Catalog No. 5000014681
Encompass_Preferred
The CloneSelect Imager is a benchtop automated imaging system for rapid brightfield and optional fluorescence imaging of mammalian cells in SBS microplates. It delivers high-resolution images and automated analysis to support clone screening, confluence measurement, and monoclonality documentation, accelerating cell line development workflows.
  • Automated brightfield and optional multichannel fluorescence imaging.
  • High-resolution CMOS imaging with standard and high-resolution modes.
  • Compatible with 6, 12, 24, 96, and 384-well SBS microplates.
  • Fast acquisition times (96-well in ~90 s for brightfield; fluorescence runs in minutes).
  • Automated colony detection, confluence measurement, and monoclonality reporting.
  • Windows-based analysis software with guided workflows for reproducible results.
Catalog No. 50-000-14681 Supplier Molecular Devices Supplier No. CLONESELECTIMAGER
Add to Cart
Add to Cart
Request a Quote
The CloneSelect Imager is a benchtop automated imaging system for rapid brightfield and optional fluorescence imaging of mammalian cells in SBS microplates. It delivers high-resolution images and automated analysis to support clone screening, confluence measurement, and monoclonality documentation, accelerating cell line development workflows.
  • Automated brightfield and optional multichannel fluorescence imaging.
  • High-resolution CMOS imaging with standard and high-resolution modes.
  • Compatible with 6, 12, 24, 96, and 384-well SBS microplates.
  • Fast acquisition times (96-well in ~90 s for brightfield; fluorescence runs in minutes).
  • Automated colony detection, confluence measurement, and monoclonality reporting.
  • Windows-based analysis software with guided workflows for reproducible results.