Learn More
Hach Company Automated Flow Cell Kit for Hach TL23 Series Turbidimeters (TL2300)
Catalog No. NC3736266 Shop All Hach Company Products
Encompass
The automated flow cell kit provides a single, dedicated flow cell for TL23-series turbidimeters to enable continuous or automated turbidity measurements, increasing throughput and simplifying sample handling.
  • Increases speed of measurement by eliminating individual sample cells.
  • Provides a single cell for all measurements, improving repeatability.
  • Eliminates the need for matched sample cells.
  • Reduces the amount of glassware that must be purchased, stored, and cleaned.
Catalog No. NC3736266 Supplier Hach Company Supplier No. 9658300
The automated flow cell kit provides a single, dedicated flow cell for TL23-series turbidimeters to enable continuous or automated turbidity measurements, increasing throughput and simplifying sample handling.
  • Increases speed of measurement by eliminating individual sample cells.
  • Provides a single cell for all measurements, improving repeatability.
  • Eliminates the need for matched sample cells.
  • Reduces the amount of glassware that must be purchased, stored, and cleaned.