Learn More
Braintree Scientific Automated Milker for Rat and Mouse
SDP
Catalog No. 501954011
Encompass
Automated Milker for Rat and Mouse - Ideal for the analysis of breast milk composition. Automated milker provides researchers the ability to collect rat/mouse milk directly from the teat. Collects milk from lactating mothers by alternating negative and atmospheric pressures through two solenoid valves and a vacuum pump attached to a microcomputer. Environmental pollutants, in particular carcinogenic s substances and endocrine disrupting chemicals, are a worldwide concern. The influence that these substances have on babies and their development, through breast or cow milk, is of great concern. The ability to gather milk from rats and mice, widely used experimental animals, will aid in advancement of research in this area. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4011 Supplier Braintree Scientific Supplier No. 59150
Add to Cart
Add to Cart
Request a Quote
Automated Milker for Rat and Mouse - Ideal for the analysis of breast milk composition. Automated milker provides researchers the ability to collect rat/mouse milk directly from the teat. Collects milk from lactating mothers by alternating negative and atmospheric pressures through two solenoid valves and a vacuum pump attached to a microcomputer. Environmental pollutants, in particular carcinogenic s substances and endocrine disrupting chemicals, are a worldwide concern. The influence that these substances have on babies and their development, through breast or cow milk, is of great concern. The ability to gather milk from rats and mice, widely used experimental animals, will aid in advancement of research in this area. **Sold for research purposes only and not for human use**