Learn More
Stoelting™ Automated Milker for Rats and Mice
SDP
Catalog No. 10001045 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Automated
Catalog No. Model
10-001-045 Automated
1 options
Catalog No. 10-001-045 Supplier Stoelting™ Supplier No. 59150
Add to Cart
Add to Cart
Request a Quote
Stoelting™ Automated Milker provides researchers the ability to collect rat/mouse milk directly from the teat.
User collects milk from lactating mothers by alternating negative and atmospheric pressures through two solenoid valves and a vacuum pump attached to a microcomputer.
Specifications
For Use With (Application) Neuroscience
Animal Mouse, Rat
Model Automated