Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arlington Scientific Automated RPR Control Set
SDP

Catalog No. 22415224 Shop All Arlington Scientific Products
Change view
Click to view available options
Quantity:
2.5 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-415-224 2.5 mL
22-415-225 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-415-224 Supplier Arlington Scientific Supplier No. 905002.5A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Consists of human serum reagents that are reactive, weak reactive and nonreactive in nontreponemal flocculation tests for the detection of reagin antibodies in human serum. These materials are intended for use by health professionals.

Specifications

Content And Storage 2°C to 8°C
Control Sets Yes
CPT Code 86592
Description Three level control for RPR
CE Marker Self-declared
Clia Complexity Moderate
Includes Reactive, weak reactive, nonreactive
Material Human Sourced
Packaging Type Kit
Quantity 2.5 mL
Test Time 7 min.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.