Learn More
RWD LIFE SCIENCE INC AUTOMATED STEREOTAXIC
SDP
Catalog No. NC3404303
Encompass_Preferred
RWD 71000 is an automated and intelligent stereotaxic instrument with high precision (1 um). The software has automatic procedures (skull window, tissue removal and automated injection procedure) and built-in rat and mouse brain atlas, which can be used to observe the position of the probe relative to the brain area. The operation of 71000 is easy-to-use, greatly reducing the errors and damage caused by manual operation. 71000 also has an anti-collision function to prevent animal damage caused by wrong movement, which is safer.
Catalog No. NC3404303 Supplier RWD LIFE SCIENCE INC Supplier No. 71000S/D/I
Add to Cart
Add to Cart
Request a Quote
RWD 71000 is an automated and intelligent stereotaxic instrument with high precision (1 um). The software has automatic procedures (skull window, tissue removal and automated injection procedure) and built-in rat and mouse brain atlas, which can be used to observe the position of the probe relative to the brain area. The operation of 71000 is easy-to-use, greatly reducing the errors and damage caused by manual operation. 71000 also has an anti-collision function to prevent animal damage caused by wrong movement, which is safer.