Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma-Aldrich Automated Wright Stain Set

Catalog No. 01288044 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Includes:
Two Reagent Wright Stains and a Rinse Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
01-288-044 Two Reagent Wright Stains and a Rinse Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-288-044 Supplier Sigma-Aldrich Supplier No. WSHT1SET
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Intended for use with Hema-Tek hematology stainers.

  • Packaging: set contains two reagent Wright Stains with a rinse solution
  • Specific packaging intended for use on the Hema-Tek hematology stainers
  • Form: solution
  • Shelf life: expiry date on the label
  • IVD: for in vitro diagnostic use
  • Applications: hematology and histology
  • Storage temperature: room temperature
TRUSTED_SUSTAINABILITY

Specifications

Includes Two Reagent Wright Stains and a Rinse Solution
Product Type Wright Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.