Learn More
Highres Biosolutions Inc AUTOMATED Z-AXIS
Supplier: Highres Biosolutions Inc A000710 R0
- Z-axis travel up to 1420 mm (55.9 in)
- Positional accuracy ±0.2 mm (0.008 in)
- Payload capacity up to 8.0 kg (17.6 lb)
- Compatible with ACell and ZCell robotic arms
- Designed for modular integration
- Compatible with Cellario control software
- Engineered for robust continuous operation
- Seamless integration into automated workflows
- Focus on lab automation, drug discovery, genomics, synthetic biology