Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Automatic Burettes

Catalog No. 10987053 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Capacity (Metric):
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
10-987-053 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-987-053 Supplier Bel-Art™ Supplier No. H379180109
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Self zeroing glass burettes.

  • With easy-to-read blue graduations against white background
  • Delivery end fitted into plastic push button stopcock which permits drop by drop delivery
  • Small screw clamp adjusts drop speed as needed for titration
  • Polyethylene filling tube extends from top of burette into bottle reservoir
  • Polyethylene plastic bottle, screw cap and round base
  • Class B graduation accuracy

Specifications

Accuracy Class B
Autoclavable Not Autoclavable
Closure Type Screw Cap
Color Natural
Diameter (Metric) Outer 7.62 cm
Capacity (Metric) 10 mL
Length (Metric) 21 cm
For Use With (Equipment) Stirplate
Material Glass
For Use With (Application) Titration, Reagent Dispensing
Includes Bottle, Round Base, Screw Cap
Scale Blue
Stopcock No
Tolerance Class B
Warranty None
Volume (Metric) Reservoir 500 mL
Type Automatic Buret
Class Class B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.